accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHTm

CAS RN 14589-44-7

IUPAC Name: thulium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: thulium(3+) triacetylacetonate
Molecular Weight (g/mol): 466.26
Molecular Formula: C15H21O6Tm
InChi Key: QTKHRYPCYVXQDE-UHFFFAOYSA-N
SMILES: [Tm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results