accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOCH3

CAS RN 14590-53-5

IUPAC Name: 2,2-dimethylcyclopropane-1-carboxylic acid
Synonyms: 2,2-dimethylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: BFNMOMYTTGHNGJ-UHFFFAOYNA-N
SMILES: CC1(C)CC1C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results