Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2,2-Dimethylcyclopropanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D36775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36775G 5 g
D36771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36775G Supplier TCI America Supplier No. D36775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14590-53-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD08460918
InChI Key BFNMOMYTTGHNGJ-SCSAIBSYSA-N
PubChem CID 10192561
IUPAC Name (1S)-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(CC1C(=O)O)C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 114.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-2,2-Dimethylcyclopropanecarboxylic Acid
EINECSNumber (3)-3774
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.