accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOCCCHHHCrCF3CF3CF3CF3CF3CF3

CAS RN 14592-80-4

IUPAC Name: chromium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Synonyms: chromium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Molecular Weight (g/mol): 673.15
Molecular Formula: C15H3CrF18O6
InChi Key: GGQIHZBUYJMHAF-UHFFFAOYSA-N
SMILES: [Cr+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results