CAS RN 14593-46-5
Sodium tert-pentyloxide, 95%
CAS: 14593-46-5 Molecular Formula: C5H11NaO Molecular Weight (g/mol): 110.132 MDL Number: MFCD00064245 InChI Key: CGRKYEALWSRNJS-UHFFFAOYSA-N Synonym: sodium tert-pentoxide,sodium 2-methylbutan-2-olate,sodium tert-amylate,sodium t-amylate,sodium-t-amylate,unii-51z39py5z8,sodium-tert-pentoxide,sodium tert-amoxide,2-butanol, 2-methyl-, sodium salt,tert-amyl alcohol sodium salt PubChem CID: 2733478 IUPAC Name: sodium;2-methylbutan-2-olate SMILES: CCC(C)(C)[O-].[Na+]
Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™
CAS: 14593-46-5 | C5H11NaO | 110.13 g/mol
Sodium tert-pentoxide, 95%, pure
CAS: 14593-46-5 Molecular Formula: C5H11NaO Molecular Weight (g/mol): 110.13 MDL Number: MFCD00064245 InChI Key: CGRKYEALWSRNJS-UHFFFAOYSA-N Synonym: sodium tert-pentoxide,sodium 2-methylbutan-2-olate,sodium tert-amylate,sodium t-amylate,sodium-t-amylate,unii-51z39py5z8,sodium-tert-pentoxide,sodium tert-amoxide,2-butanol, 2-methyl-, sodium salt,tert-amyl alcohol sodium salt PubChem CID: 2733478 IUPAC Name: sodium;2-methylbutan-2-olate SMILES: CCC(C)(C)[O-].[Na+]
Sodium tert-Pentoxide 97.0+%, TCI America™
CAS: 14593-46-5 Molecular Formula: C5H11NaO Molecular Weight (g/mol): 110.132 MDL Number: MFCD00064245 InChI Key: CGRKYEALWSRNJS-UHFFFAOYSA-N Synonym: sodium tert-pentoxide,sodium 2-methylbutan-2-olate,sodium tert-amylate,sodium t-amylate,sodium-t-amylate,unii-51z39py5z8,sodium-tert-pentoxide,sodium tert-amoxide,2-butanol, 2-methyl-, sodium salt,tert-amyl alcohol sodium salt PubChem CID: 2733478 IUPAC Name: sodium;2-methylbutan-2-olate SMILES: CCC(C)(C)[O-].[Na+]
Sigma Aldrich Sodium Tert-Pentoxide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Sodium Tert-Pentoxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Sodium Tert-Pentoxide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.