Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tert-Pentoxide

Catalog No. 280704500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-070-4500G 500 g
28-070-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 28-070-4500G Supplier Sigma Aldrich Supplier No. 280704500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tert-pentoxide (NaOt-Am) is a commonly used base in the deoxycyanamidation of alcohols and in the N-cyanation of secondary amines. It can also be used in the preparation of complex reducing agents to reduce 1-halonaphthalenes and substituted halobenzenes to corresponding naphthalenes and benzenes.

Specifications

CAS 14593-46-5
Molecular Formula C5H11NaO
Linear Formula NaOC(CH3)2CH2CH3
MDL Number MFCD00064245
Quantity 500 g
Synonym NaOt-Am; tert-Amyl alcohol sodium salt; Sodium tert-amoxide
Molecular Weight (g/mol) 110.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.