accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SH3CCH3FPFFFFF

CAS RN 146062-15-9

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tris(4-methylphenyl)sulfanium
Synonyms: tris(4-methylphenyl)sulfanium hexafluorophosphate
Molecular Weight (g/mol): 450.42
Molecular Formula: C21H21F6PS
InChi Key: BIWXKHOYLGAZDG-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CC1=CC=C(C=C1)[S+](C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results