Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-p-tolylsulfonium Hexafluorophosphate 90.0+%, TCI America™
SDP

Catalog No. T20411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20411G Supplier TCI America Supplier No. T20411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146062-15-9
Molecular Formula C21H21F6PS
Molecular Weight (g/mol) 450.42
MDL Number MFCD06797187
InChI Key BIWXKHOYLGAZDG-UHFFFAOYSA-N
Synonym Tris(4-methylphenyl)sulfonium Hexafluorophosphate
PubChem CID 13750251
IUPAC Name hexafluoro-λ⁵-phosphanuide; tris(4-methylphenyl)sulfanium
SMILES F[P-](F)(F)(F)(F)F.CC1=CC=C(C=C1)[S+](C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 171°C
Color White
Quantity 1 g
Formula Weight 450.42
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tri-p-tolylsulfonium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.