accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNOONHNOOHOOOO(E/Z)

CAS RN 14611-34-8

IUPAC Name: 5-{[amino({[(benzyloxy)carbonyl]imino})methyl][(benzyloxy)carbonyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino({[(benzyloxy)carbonyl]imino})methyl][(benzyloxy)carbonyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 576.61
Molecular Formula: C30H32N4O8
InChi Key: YSGAXJCIEJGVFV-UHFFFAOYNA-N
SMILES: NC(=NC(=O)OCC1=CC=CC=C1)N(CCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)