Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(carbobenzoxy)-L-arginine 98.0+%, TCI America™
SDP

Catalog No. T07551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07551G Supplier TCI America Supplier No. T07551G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14611-34-8
Molecular Formula C30H32N4O8
Molecular Weight (g/mol) 576.606
MDL Number MFCD00038693
InChI Key YSGAXJCIEJGVFV-RUZDIDTESA-N
Synonym Tri-Cbz-L-arginine, Z-Arg(Z)2-OH
PubChem CID 131843446
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)-5-[phenylmethoxycarbonyl-[(E)-N'-phenylmethoxycarbonylcarbamimidoyl]amino]pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN(C(=NC(=O)OCC2=CC=CC=C2)N)C(=O)OCC3=CC=CC=C3)C(=O)O

Specifications

Melting Point 135°C
Color White
Quantity 1 g
Formula Weight 576.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(carbobenzoxy)-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.