accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14613-37-7

IUPAC Name: 1-(1-methylpiperidin-3-yl)methanamine
Synonyms: 1-(1-methylpiperidin-3-yl)methanamine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: KEDTYNCWGSIWBK-UHFFFAOYNA-N
SMILES: CN1CCCC(CN)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results