accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 1462-33-5

IUPAC Name: 1-chloro-2-(chloromethoxy)ethane
Synonyms: 1-chloro-2-(chloromethoxy)ethane
Molecular Weight (g/mol): 128.98
Molecular Formula: C3H6Cl2O
InChi Key: LUTWEKBTDWRTSE-UHFFFAOYSA-N
SMILES: ClCCOCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results