accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N(R)O(R)HO(S)HO(R)HOOHNO

CAS RN 1463-10-1

IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: thymidin thymine riboside ribosylthymine ribosylthymidine β-delta-ribofuranoside thymine ribonucleoside 1-β-delta-ribofuranosylthymine thymine ribofuranoside β-D-ribofuranoside thymine-1 ribothymidine
Molecular Weight (g/mol): 258.23
Molecular Formula: C10H14N2O6
InChi Key: DWRXFEITVBNRMK-JXOAFFINSA-N
SMILES: CC1=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)NC1=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)