accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNOHN

CAS RN 14631-20-0

IUPAC Name: N-(2-oxo-2,3-dihydropyrimidin-4-yl)acetamide
Synonyms: N-(2-oxo-3H-pyrimidin-4-yl)acetamide
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: IJCKBIINTQEGLY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=NC(=O)N1
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)