accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146328-85-0

IUPAC Name: 5-bromo-2-fluorobenzoate
Synonyms: 5-bromo-2-fluorobenzoate
Molecular Weight (g/mol): 218
Molecular Formula: C7H3BrFO2
InChi Key: PEXAZYDITWXYNJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Br)=CC=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results