Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluorobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. B29745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29745G 5 g
B29741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29745G Supplier TCI America Supplier No. B29745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146328-85-0
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00143423
InChI Key PEXAZYDITWXYNJ-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
PubChem CID 2736315
IUPAC Name 5-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)F

Specifications

Melting Point 150°C
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.