accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14634-91-4

IUPAC Name: λ²-iron(2+) tris(1,10-phenanthroline) sulfate
Synonyms: ferroin
Molecular Weight (g/mol): 692.53
Molecular Formula: C36H24FeN6O4S
InChi Key: CIWXFRVOSDNDJZ-UHFFFAOYSA-L
SMILES: [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results