Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Tris)-1,10-Phenanthroline Iron (II) Sulfate Solution (0.025M/Certified), Fisher Chemical™

Catalog No. P69500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P69-500 500 mL Glass Bottle
P69-100 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P69-500

missing translation for 'validMainframeMsg'

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14634-91-4 , 7782-63-0
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 84567
IUPAC Name λ²-iron(2+) tris(1,10-phenanthroline) sulfate
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Color Red
Melting Point 0°C
Linear Formula (C12H8N2)3FeSO4
Density 1.0g/cm³
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Suitability Pass Test (redox indicator)
Vapor Pressure 14mmHg
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material (Tris)-1, 10-Phenanthroline Iron (II) Sulfate Solution, 0.025M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.