accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146346-81-8

SiOOOHOONHH3CCH3H3CCH3H3C(S)

CAS RN 146346-81-8

IUPAC Name: (2S)-3-[(tert-butyldimethylsilyl)oxy]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-[(tert-butyldimethylsilyl)oxy]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 441.6
Molecular Formula: C24H31NO5Si
InChi Key: IONOZYFSQPGBAT-NRFANRHFSA-N
SMILES: CC(C)(C)[Si](C)(C)OC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)