accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14635-75-7

IUPAC Name: dioxoamine; tetrafluoroboranuide
Synonyms: nitrite ion tetrafluoroborate
Molecular Weight (g/mol): 133.82
Molecular Formula: BF4HNO2
InChi Key: XYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES: [O-][NH+]=O.F[B-](F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)