Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nitrosyl Tetrafluoroborate

Catalog No. 1750645G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-506-45G 5 g
17-506-450G 50 g
17-506-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 17-506-45G Supplier Sigma Aldrich Supplier No. 1750645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nitrosyl tetrafluoroborate is an efficient nitrosating and diazotizing agent. It reacts with alcohols and secondary amines to yield alkyl nitrites and nitrosamines, respectively. It reacts with primary amines to yield diazonium tetrafluoroborates. NOBF4 is also a mild oxidant and commonly used for single electron transfer oxidation.

Specifications

CAS 14635-75-7
Quantity 5 g
Molecular Formula BF4NO
Linear Formula NOBF4
MDL Number MFCD00011433
Synonym Nitrosonium tetrafluoroborate
Molecular Weight (g/mol) 116.81 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.