accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14637-34-4

IUPAC Name: ammonium tetraphenylboranuide
Synonyms: ammonium tetraphenylborate
Molecular Weight (g/mol): 337.27
Molecular Formula: C24H24BN
InChi Key: ZWFYDLYRTYMBIL-UHFFFAOYSA-O
SMILES: [NH4+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)