accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14638-70-1

IUPAC Name: 2,3-dihydroxybutanedioic acid; 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol
Synonyms: (.+-.)-tartaric acid; isoproterenol
Molecular Weight (g/mol): 361.35
Molecular Formula: C15H23NO9
InChi Key: LBOPECYONBDFEM-UHFFFAOYNA-N
SMILES: OC(C(O)C(O)=O)C(O)=O.CC(C)NCC(O)C1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results