Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Isoproterenol L(+)-bitartrate, Thermo Scientific™

Catalog No. AC182840010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC182840010 Supplier Thermo Scientific Chemicals Supplier No. 182840010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14638-70-1
Molecular Formula C15H23NO9
Molecular Weight (g/mol) 361.347
InChI Key LBOPECYONBDFEM-VUAFVNRWSA-N
Synonym s-4-1-hydroxy-2-isopropylamino ethyl benzene-1,2-diol 2r,3r-2,3-dihydroxysuccinate, d-isoproterenol bitartrate, +-isoprenaline bitartrate, s-isoproterenol bitartrate, +-isoproterenol bitartrate, l-+-isoproterenol bitartrate, d-n-isopropylnorepinephrine bitartrate, l +-tartaric acid; l-isoproterenol, benzyl alcohol, 3,4-dihydroxy-alpha-isopropylamino methyl-, +-, +-tartrate 1:1 salt, prestwick3_000911
PubChem CID 2724018
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;4-[(1S)-1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol
SMILES CC(C)NCC(C1=CC(=C(C=C1)O)O)O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 169°C to 175°C
Color White to Beige
Infrared Spectrum Authentic
Quantity 1 g
Specific Rotation +34° to +42° (20°C, 589nm) (c=1, H2O)
Formula Weight 361.34
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material (S)-(+)-Isoproterenol L(+)-bitartrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.