accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NO

CAS RN 1465-76-5

IUPAC Name: 1-tert-butylpiperidin-4-one
Synonyms: 1-tert-butylpiperidin-4-one
Molecular Weight (g/mol): 155.24
Molecular Formula: C9H17NO
InChi Key: NKPMJJGLSFUOPK-UHFFFAOYSA-N
SMILES: CC(C)(C)N1CCC(=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results