accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(R)NOO

CAS RN 146727-62-0

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-cyanophenyl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(4-cyanophenyl)propanoate
Molecular Weight (g/mol): 289.31
Molecular Formula: C15H17N2O4
InChi Key: RMBLTLXJGNILPG-GFCCVEGCSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results