Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-4-cyano-D-phenylalanine, 98%

Catalog No. AAH62947MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62947MD 250 mg
AAH6294703 1 g
AAH6294706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62947MD Supplier Thermo Scientific Chemicals Supplier No. H62947MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 146727-62-0 | C15H17N2O4 | 289.31 g/mol

N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146727-62-0
Molecular Formula C15H17N2O4
Molecular Weight (g/mol) 289.31
MDL Number MFCD00672527
InChI Key RMBLTLXJGNILPG-GFCCVEGCSA-M
Synonym boc-d-phe 4-cn-oh, r-n-boc-4-cyanophenylalanine, boc-4-cyano-d-phenylalanine, boc-d-4-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, d-phenylalanine, 4-cyano-n-1,1-dimethylethoxy carbonyl, boc-d-4-cyanophe, boc-4-cyano-d-phe-oh, boc-d-4-cn-phe-oh
PubChem CID 2734494
IUPAC Name (2R)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O

Specifications

Quantity 250 mg
UN Number UN3439
Beilstein 9339089
Formula Weight 290.31
Percent Purity 98%
Chemical Name or Material N-Boc-4-cyano-D-phenylalanine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.