accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14676-00-7

OHOOOHNH3CCH3CH3H3C

CAS RN 14676-00-7

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}octanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]octanoic acid
Molecular Weight (g/mol): 259.35
Molecular Formula: C13H25NO4
InChi Key: PGJIJHNIIQRRGV-UHFFFAOYSA-N
SMILES: CCCCCCC(NC(=O)OC(C)(C)C)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)