accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 146885-81-6

IUPAC Name: 3,8-dimethyl-1,2-dihydroacenaphthylene-1,2-dione
Synonyms: 3,8-dimethylacenaphthylene-1,2-dione
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: YMVJXPYFESLYEO-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC(C)=C3C(=O)C(=O)C1=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)