Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,8-Dimethylacenaphthenequinone 97.0+%, TCI America™
SDP

Catalog No. D5156200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5156200MG 200 mg
D51561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5156200MG Supplier TCI America Supplier No. D5156200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146885-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
InChI Key YMVJXPYFESLYEO-UHFFFAOYSA-N
Synonym 3,8-Dimethylacenaphthylene-1,2-dione
PubChem CID 11074654
IUPAC Name 3,8-dimethylacenaphthylene-1,2-dione
SMILES CC1=C2C3=C(C=C1)C=CC(=C3C(=O)C2=O)C

Specifications

Melting Point 208°C
Quantity 200 mg
Formula Weight 210.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,8-Dimethylacenaphthenequinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.