CAS RN 146986-50-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Y-27632
Y-27632A, CAS 146986-50-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.
2
MilliporeSigma™ Calbiochem™ InSolution™ Y-27632, Sterile-Filtered, Cell Culture-Tested
A 10 mM (2 mg/591 μL) sterile-filtered solution of Y-27632 in H2O.