accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146987-10-2

IUPAC Name: (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl]pentanamido}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid|Fmoc-Lys(Biotin)-OH
Molecular Weight (g/mol): 594.73
Molecular Formula: C31H38N4O6S
InChi Key: OFIBQNGDYNGUEZ-CVKYCUBCNA-N
SMILES: [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)