Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine 95.0+%, TCI America™
SDP

Catalog No. F1042200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1042200MG 200 mg
F10421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1042200MG Supplier TCI America Supplier No. F1042200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 146987-10-2 | C31H38N4O6S | 594.73 g/mol

Chemical Identifiers

CAS 146987-10-2
Molecular Formula C31H38N4O6S
Molecular Weight (g/mol) 594.73
MDL Number MFCD00270741
InChI Key OFIBQNGDYNGUEZ-CVKYCUBCNA-N
Synonym fmoc-lys biotin-oh, fmoc-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamido hexanoic acid, fmoc biocytin, fmoc-biocytin, fmoc-lys biotin, n, a-fmoc-biocytin, fmoc-l-lys biotinyl-oh
PubChem CID 11039242
IUPAC Name (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2

Specifications

Melting Point 183°C
Color White
Quantity 200 mg
Formula Weight 594.73
Percent Purity ≥95.0% (HPLC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.