accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1470-57-1

CH3OHO

CAS RN 1470-57-1

IUPAC Name: 2-benzoyl-4-methylphenol
Synonyms: 2-benzoyl-4-methylphenol
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: OQERFUGURPLBQH-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results