Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-methylbenzophenone 97.0+%, TCI America™
SDP

Catalog No. H10085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10085G 5 g
H100825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10085G Supplier TCI America Supplier No. H10085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1470-57-1
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00002379
InChI Key OQERFUGURPLBQH-UHFFFAOYSA-N
Synonym 2-hydroxy-5-methylbenzophenone, 2-benzoyl-4-methylphenol, 2-hydroxy-5-methylphenyl phenyl methanone, unii-85w615bmp7, methanone, 2-hydroxy-5-methylphenyl phenyl, 2-hydroxy-5-methyl-benzophenone, 2-hydroxy-5-methylphenyl-phenylmethanone, acmc-209cy0, dsstox_cid_27857, dsstox_rid_82610
PubChem CID 73851
IUPAC Name (2-hydroxy-5-methylphenyl)-phenylmethanone
SMILES CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 212.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-methylbenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.