accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOOH

CAS RN 1470-79-7

IUPAC Name: 4-(4-hydroxybenzoyl)benzene-1,3-diol
Synonyms: 4-(4-hydroxybenzoyl)benzene-1,3-diol
Molecular Weight (g/mol): 230.22
Molecular Formula: C13H10O4
InChi Key: OKJFKPFBSPZTAH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C1=CC=C(O)C=C1O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)