Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,4'-Trihydroxybenzophenone, Thermo Scientific™

Catalog No. AC139810250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC139810250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139810250 Supplier Thermo Scientific Chemicals Supplier No. 139810250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1470-79-7
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C13H10O4
Linear Formula (HO)2C6H3COC6H4OH
Quantity 25g
MDL Number MFCD00002356
Beilstein 08, 422
Synonym 2,4,4'-trihydroxybenzophenone, 2,4-dihydroxyphenyl 4-hydroxyphenyl methanone, benzophenone, 2,4,4'-trihydroxy, unii-qp9121ig2s, methanone, 2,4-dihydroxyphenyl 4-hydroxyphenyl, 2,4-dihydroxyphenyl p-hydroxybenzyl ketone, 2,4-dihydroxyphenyl-4-hydroxyphenyl methanone, 2,4-dihydroxy-phenyl-4-hydroxy-phenyl-methanone, dsstox_cid_24836, dsstox_rid_80517
InChI Key OKJFKPFBSPZTAH-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=C(O)C=C1O
IUPAC Name 4-(4-hydroxybenzoyl)benzene-1,3-diol
Molecular Weight (g/mol) 230.22
PubChem CID 73852
Formula Weight 230.22
Percent Purity ≥94%
Chemical Name or Material 2, 4, 4'-Trihydroxybenzophenone
Show More Show Less
EINECSNumber 216-004-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.