CAS RN 147098-18-8
IUPAC Name:
sodium 7-[4-(4-fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Synonyms:
sodium 7-[4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethanesulfonamido)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol):
503.52
Molecular Formula:
C22H27FN3NaO6S
InChi Key:
RGEBGDYYHAFODH-UHFFFAOYNA-M
SMILES:
[Na+].CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1C=CC(O)CC(O)CC([O-])=O)N(C)S(C)(=O)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Rosuvastatin, sodium salt, 97%
Thermo Scientific Chemicals Rosuvastatin, sodium salt, 97%
CAS: 147098-18-8 Molecular Formula: C22H27FN3NaO6S Molecular Weight (g/mol): 503.53 InChI Key: RGEBGDYYHAFODH-GBNZRNLASA-M Synonym: rosuvastatin sodium PubChem CID: 131632367 IUPAC Name: sodium;(3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate SMILES: CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)[O-])O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Na+]