Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rosuvastatin, sodium salt, 97%

Catalog No. AC461490010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC461490010 Supplier Thermo Scientific Chemicals Supplier No. 461490010
  • $260.00
    $221.65 / Each of 1

    Save $38.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 147098-18-8 | C22H27FN3NaO6S | 503.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147098-18-8
Molecular Formula C22H27FN3NaO6S
Molecular Weight (g/mol) 503.53
InChI Key RGEBGDYYHAFODH-GBNZRNLASA-M
Synonym rosuvastatin sodium
PubChem CID 131632367
IUPAC Name sodium;(3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
SMILES CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)[O-])O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Na+]

Specifications

Melting Point 106.0°C to 118.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 503.53
Percent Purity 98%
Chemical Name or Material Rosuvastatin, sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.