accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNOFNCH3SOOH3C

CAS RN 147118-37-4

IUPAC Name: N-[4-(4-fluorophenyl)-5-formyl-6-(propan-2-yl)pyrimidin-2-yl]-N-methylmethanesulfonamide
Synonyms: N-[4-(4-fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl]-N-methylmethanesulfonamide
Molecular Weight (g/mol): 351.4
Molecular Formula: C16H18FN3O3S
InChi Key: WOCOTUDOVSLFOB-UHFFFAOYSA-N
SMILES: CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1C=O)N(C)S(C)(=O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)