accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNH2OH2N

CAS RN 147123-47-5

IUPAC Name: 3-aminothiophene-2-carboxamide
Synonyms: 3-aminothiophene-2-carboxamide
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: BKDZTJNNXCNSCK-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)C=CS1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results