Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminothiophene-2-carboxamide, 97%, Thermo Scientific™

Catalog No. AAL1237103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1237103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1237103 Supplier Thermo Scientific Chemicals Supplier No. L1237103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 147123-47-5
Melting Point 120°C to 122°C
Molecular Formula C5H6N2OS
Quantity 1 g
MDL Number MFCD00052593
Beilstein 7165908
Synonym 3-amino-2-thiophenecarboxamide, 2-thiophenecarboxamide, 3-amino, 3-amino-2-carbamoylthiophene, 3-amino-thiophene-2-carboxylic acid amide, pubchem5242, acmc-20aa6u, 3-azanylthiophene-2-carboxamide, 3-amino-thiophene-2-carboxamide, 3-amino-2-aminocarbonyl-thiophene, 3-amino-2-thiophenecarboxamide #
InChI Key BKDZTJNNXCNSCK-UHFFFAOYSA-N
SMILES C1=CSC(=C1N)C(=O)N
IUPAC Name 3-aminothiophene-2-carboxamide
Molecular Weight (g/mol) 142.176
PubChem CID 518993
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material 3-Aminothiophene-2-carboxamide
Show More Show Less
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.