accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1474-78-8

IUPAC Name: ethyl (diethoxyphosphoryl)formate
Synonyms: ethyl (diethoxyphosphoryl)formate
Molecular Weight (g/mol): 210.17
Molecular Formula: C7H15O5P
InChi Key: NOJFJZZMRDSOLM-UHFFFAOYSA-N
SMILES: CCOC(=O)P(=O)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results