Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Diethoxyphosphinylformate, TCI America™
SDP

Catalog No. E040025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E040025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E040025G Supplier TCI America Supplier No. E040025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1474-78-8
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.166
MDL Number MFCD00009108
InChI Key NOJFJZZMRDSOLM-UHFFFAOYSA-N
Synonym ethyl diethoxyphosphinylformate, triethyl phosphonoformate, triethyl carboxyphosphonate, diethyl ethoxycarbonylphosphonate, triethyl phosphonomethanoate, diethyl carbethoxyphosphonate, ethyldiethoxyphosphinylformate, ethyl diethoxyphosphoryl formate, unii-tux6m928id, phosphinecarboxylic acid, diethoxy-, ethyl ester, oxide
PubChem CID 65081
IUPAC Name ethyl diethoxyphosphorylformate
SMILES CCOC(=O)P(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 133°C
Quantity 25 g
Formula Weight 210.17
Physical Form Liquid
Chemical Name or Material Ethyl Diethoxyphosphinylformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.