accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 147404-69-1

IUPAC Name: 4'-methyl-[1,1'-biphenyl]-3-carboxylate
Synonyms: 4'-methyl-[1,1'-biphenyl]-3-carboxylate
Molecular Weight (g/mol): 211.24
Molecular Formula: C14H11O2
InChi Key: APYJGQZXCGXMGB-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)C1=CC=CC(=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results