accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1475-13-4

IUPAC Name: 1-(2,4-dichlorophenyl)ethan-1-ol
Synonyms: 1-(2,4-dichlorophenyl)ethanol
Molecular Weight (g/mol): 191.05
Molecular Formula: C8H8Cl2O
InChi Key: KWZDYNBHZMQRLS-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results