Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-alpha-methylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D45085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45085G 5 g
D450825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45085G Supplier TCI America Supplier No. D45085G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1475-13-4
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.051
MDL Number MFCD00004511
InChI Key KWZDYNBHZMQRLS-UHFFFAOYSA-N
Synonym 1-2,4-dichlorophenyl ethanol, 1-2,4-dichlorophenyl ethan-1-ol, 2,4-dichloro-alpha-methylbenzyl alcohol, 1-2,4-dichloro-phenyl ethanol, 1-2,4-dichlorophenyl ethyl alcohol, 2,4-dichloro-alpha-methylbenzylalcohol, 2,4-dichloro-1-1-hydroxyethyl benzene, 2,4-dichloro-alpha-methylbenzenemethanol, 2,4-dichloro-.alpha.-methylbenzyl alcohol, 2,4-dichloro-a-methylbenzyl alcohol
PubChem CID 92889
IUPAC Name 1-(2,4-dichlorophenyl)ethanol
SMILES CC(C1=C(C=C(C=C1)Cl)Cl)O

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
Formula Weight 191.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-alpha-methylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.