accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14752-61-5

IUPAC Name: 1-iodopent-1-yne
Synonyms: 1-iodopent-1-yne
Molecular Weight (g/mol): 194.02
Molecular Formula: C5H7I
InChi Key: UWPKZGBPLFRGAH-UHFFFAOYSA-N
SMILES: CCCC#CI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results