Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentynyl Iodide 97.0+%, TCI America™
SDP

Catalog No. P10551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10551G Supplier TCI America Supplier No. P10551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14752-61-5
Molecular Formula C5H7I
Molecular Weight (g/mol) 194.015
MDL Number MFCD00059098
InChI Key UWPKZGBPLFRGAH-UHFFFAOYSA-N
Synonym 1-Iodo-1-pentyne
PubChem CID 12643190
IUPAC Name 1-iodopent-1-yne
SMILES CCCC#CI

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
UN Number 1993
Formula Weight 194.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentynyl Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.