accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNOOCH3H3CCH3

CAS RN 147539-41-1

IUPAC Name: tert-butyl 4-(methylamino)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(methylamino)piperidine-1-carboxylate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: CZYUGTLMFHDODF-UHFFFAOYSA-N
SMILES: CNC1CCN(CC1)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)